missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Diffusion Tubes with Direct Indication

Catalog No. 17985353F
Change view
Click to view available options
Measurement Range:
0.13 to 3.8%
2.5 to 188
Humidity Range:
1 to 15mg/L
1 to 16mg/L
Temperature Range (Metric):
0°C to 40°C
10°C to 30°C
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Humidity Range Temperature Range (Metric)
17985353F 0.13 to 3.8% 1 to 15mg/L 10°C to 30°C
17985353C 2.5 to 188 1 to 16mg/L 0°C to 40°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17985353F Supplier Dräger™ Supplier No. 8101051
Only null left

Convenient, long-term TWA monitoring—direct reading, with no lab analysis required

Tubes sample ambient air by gaseous diffusion with no pumps or moving parts. To use, break off the tube end with the special holder, put the tube in the holder, and clip it to your pocket or lapel.

  • Obtain time-weighted average (TWA) measurements of exposure to toxic gases for periods of up to 8 hours
  • Reagent changes color as it reacts with the airborne contaminant
  • Final results are calculated by dividing the tube reading by the number of hours it was used
  • Designed and manufactured using ISO 90001:2000-certified processes

Order Info

Use with accessory clip-on holder (Cat. No. 17-986-4; available separately below). Ten tubes per pack.

† Higher concentration measurements possible with reduced sampling time.

Specifications

Humidity Range 1 to 15mg/L
Measurement Range 0.13 to 3.8%
Quantity 10
Response Time 8 hr.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C