Learn More
Dräger™ CMS Chips: Petroleum Hydrocarbons
Catalog No. 19038778
Change view
Click to view available options
Measurement Range:
20 to 500
Response Time:
150 to 330 sec.
Catalog No. Measurement Range Response Time
19038778 20 to 500 150 to 330 sec.
1 options
Catalog No. 19038778 Supplier Dräger™ Supplier No. 6406200
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Petroleum Hydrocarbons CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 20 to 500
Response Time 150 to 330 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C