Learn More
Dräger™ CMS Chips: Phosphine
Catalog No. 19041692
Change view
Click to view available options
Measurement Range:
0.1 to 2.5
Response Time:
25 to 350 sec.
Catalog No. Measurement Range Response Time
19041692 0.1 to 2.5 25 to 350 sec.
1 options
Catalog No. 19041692 Supplier Dräger™ Supplier No. 6406400
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Suitable for 10 measurements of one gas.
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Phosphine CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 0.1 to 2.5
Response Time 25 to 350 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C