Learn More
Dräger™ CMS Chips: Hydrochloric Acid
Catalog No. 189991923
Change view
Click to view available options
Measurement Range:
1 to 25
20 to 500
Response Time:
15 to 110 sec.
6 to 80 sec.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Response Time
189991923 1 to 25 15 to 110 sec.
189991924 20 to 500 6 to 80 sec.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 189991923 Supplier Dräger™ Supplier No. 6406090
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Hydrochloric Acid CMS Chip
Humidity Range 1 to 10mg/L
Measurement Range 1 to 25
Response Time 15 to 110 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C