Learn More
Dräger™ Civil Defense Simultest Kit: Test Sets
Catalog No. 19046636
Change view
Click to view available options
Type:
CDS Test Set I
CDS Test Set V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
19046636 CDS Test Set V
19046635 CDS Test Set I
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
No other type of tube should be substituted for use with these kits.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Catalog No. 19046636 Supplier Dräger™ Supplier No. 8103200
No other type of tube should be substituted for use with these kits.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
For use with Civil Defense Simultest Kit (Cat. No. 19-038-865)
For Chemical Warfare Agent (CWA) detection. test sets detect eight chemical agents, including nerve, blood, lung, nose, and throat irritants. Include five colorimetric DraegerTubes™, the same field-proven tube chemistry used by the North Atlantic Treaty Organization (NATO) in single detector tubes. Each test set provides on-the-spot results in less than five minutes.
CDS Test Set I detector tubes test for hydrocyanic acid, phosgene, organic arsenic compounds and arsine, organic basic nitrogen compounds, and thioether
CDS Test Set V detector tube test for phosphoric acid esters, phosgene, cyanogen chloride, chlorine, and thioether
Specifications
Type CDS Test Set V
For Use With (Application) Gas detection, no calibration
Includes Includes colorimetric detector tubes for phosphoric acid esters, phosgene, cyanogen chloride, chlorine, and thioether