missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Calibration Gases

Supplier:  Dräger™ 4594943

 View more versions of this product

Catalog No. 19038746


Only null left

Description

Description

Specifications

Chemical Identifiers

Calibration Gas

Specifications

15.32 gal.
LEL, methane 50%, carbon monoxide 100ppm, hydrogen sulfide 25ppm, oxygen 20.9%
Calibration Gas
58 L
Methane, Carbon Monoxide, Hydrogen Sulfide, Oxygen
500 psi
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

SDS
Documents

Documents

Promotions

Promotions