missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Butadiene (OV) XXS Replacement Sensor

Catalog No. 17980810
Change view
Click to view available options
Components Measured:
Butadiene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17980810 Butadiene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17980810 Supplier Dräger™ Supplier No. 6811530

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

XXS Replacement Sensor, for use with Pac 7000 and X-am 5000 multi-gas detectors.

  • Measuring range: 0 to 100 ppm
  • Resolution: 1 ppm
  • Butadiene (OV)

Specifications

Components Measured Butadiene
For Use With (Equipment) Pac 7000, X-am 5000 Multigas Detectors
Type Replacement Sensor