Learn More
Dräger™ 7000 Tube Opener
Catalog No. 296358544
Change view
Click to view available options
For Use With (Equipment):
Detector Tube Systems
Catalog No. For Use With (Equipment)
296358544 Detector Tube Systems
1 options
Catalog No. 296358544 Supplier Dräger™ Supplier No. 6401200
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
An accessory for detector tube systems.
  • Accessory for detector tube systems
  • For scoring and breaking tips off individual Draeger detector tubes
  • Glass tips safely stored for proper disposal
  • Aids in breaking tube ampoules
Specifications
For Use With (Equipment) Detector Tube Systems
Type 7000 Tube Opener