missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPP3 Recombinant Rabbit Monoclonal Antibody (23GB5190)
GREENER_CHOICE

Catalog No. MA553860
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553860 100 μL
MA553861 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553860 Supplier Invitrogen™ Supplier No. MA553860
Only null left

Rabbit Recombinant Monoclonal Antibody

DPP3 Recombinant Monoclonal Antibody for Flow, Western Blot

This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized.

Specifications

Antigen DPP3
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone 23GB5190
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DPP3
Gene Accession No. O55096, Q9NY33
Gene Alias 4930533O14Rik; C86324; Dipeptidyl aminopeptidase III; dipeptidyl arylamidase III; Dipeptidyl peptidase 3; dipeptidyl peptidase III; dipeptidylpeptidase 3; dipeptidyl-peptidase 3; dipeptidylpeptidase III; dipeptidyl-peptidase III; DPP III; Dpp3; DPPIII; Enkephalinase B; hCG_2016942
Gene Symbols DPP3
Host Species Rabbit
Immunogen A synthesized peptide derived from human DPP3 (700-737AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10072, 114591
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less