missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPEP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552984 Supplier Invitrogen™ Supplier No. PA552984
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Immunogen sequence: SLIGVEGGHS IDSSLGVLRA LYQLGMRYLT LTHSCNTPWA DNWLVDTGDS EPQSQGLSPF GQRVVKELNR LGVLIDLAHV SVATMKATLQ LSRAPVIFSH SSAYSVCASR RN Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 82%.

The protein encoded by this gene is a kidney membrane enzyme involved in the metabolism of glutathione and other similar proteins by dipeptide hydrolysis. The encoded protein is known to regulate leukotriene activity by catalyzing the conversion of leukotriene D4 to leukotriene E4. This protein uses zinc as a cofactor and acts as a disulfide-linked homodimer. Two transcript variants encoding the same protein have been found for this gene.

Specifications

Antigen DPEP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DPEP1
Gene Accession No. P16444
Gene Alias AI327012; Dehydropeptidase-I; dipeptidase 1; dipeptidase 1 (renal); Dpep1; hRDP; MBD; Mbd1; MBD-1; MDP; membrane bound dipeptidase type II; membrane-bound dipeptidase 1; Microsomal dipeptidase; RDP; Renal dipeptidase; testicular tissue protein Li 57
Gene Symbols DPEP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DPEP1. Recombinant protein control fragment (Product #RP-89644).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1800
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less