missing translation for 'onlineSavingsMsg'
Learn More

Doxylamine Succinate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014625
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014625 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014625 Supplier Merck Emd Millipore Supplier No. PHR14201G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Doxylamine succinate is an antihistaminic drug generally employed for the prevention of morning sickness in pregnant women. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 562-10-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C17H22N2 O · C4H6O4
MDL Number MFCD00056168
Synonym Doxylamine succinate salt
Type Certified Reference Material
Quantity 1 g
Formula Weight 388.46
Grade Certified Reference Material