missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxycycline Hyclate USP, Medisca™

$138.00 - $11,647.85
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
18048041
![]() |
N/A | 25 g |
Each for $138.00
|
|
|||||
18048101
![]() |
N/A | 5 kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18048102
![]() |
N/A | 100 g |
Each for $426.30
|
|
|||||
18048118
![]() |
N/A | 1 kg |
Each for $2,793.88
|
|
|||||
Description
Doxycycline Hyclate USP is a yellow crystalline powder. It is hygroscopic.
Chemical Identifiers
| 24390-14-5 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 24390-14-5 | |
| 800 to 920 μg/mg (Doxycycline) |
| Crystalline Powder | |
| Yellow | |
| USP |