Learn More
Doxycycline Hyclate USP, Medisca™
Supplier: Medisca Pharmaceutique Inc 043403
| Quantity | 5 kg |
|---|
Description
Doxycycline Hyclate USP is a yellow crystalline powder. It is hygroscopic.
Chemical Identifiers
| 24390-14-5 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 24390-14-5 | |
| 5 kg | |
| USP |
| Crystalline Powder | |
| Yellow | |
| 800 to 920 μg/mg (Doxycycline) |