missing translation for 'onlineSavingsMsg'
Learn More

Doxepin hydrochloride

Potent H1-histamine receptor antagonist and a 5-HT receptor antagonist | CAS: 1229-29-4 | C19H22ClNO | 315.841 g/mol

Supplier:  Thermo Scientific Chemicals J6257903

 View more versions of this product

Catalog No. AAJ6257903

Only null left
Explore available promotions

Description

Description

It is a highly potent H1 histamine receptor antagonist (Kd = 310 pM) and tricyclic antidepressant that inhibits the reuptake of serotonin and norepinephrine and acts as an antagonist at histamine, serotonin, adrenergic, and muscarinic receptors with Ki values in the nanomolar range. Also binds to the H4 histamine receptor (pKi = 6.79). Used clinically to treat anxiety and depression.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (100 mM), chloroform (1:2), alcohol (1:1), methanol, and DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Doxepin hydrochloride
187°C to 189°C
C19H22ClNO
1 g
14,3435
Soluble in water (100 mM),chloroform (1:2),alcohol (1:1),methanol,and DMSO.
CN(C)CCC=C1C2=CC=CC=C2COC3=CC=CC=C31.Cl
315.841
315.84
1229-29-4
White
MFCD00079135
UN2811
doxepin hydrochloride, novoxapin, aponal, toruan, curatin, sinequan, zonalon, adapin, doxepine hydrochloride, novoxapin hydrochloride
MHNSPTUQQIYJOT-SJDTYFKWSA-N
(3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
6419921
Powder

Chemical Identifiers

1229-29-4
315.841
MHNSPTUQQIYJOT-SJDTYFKWSA-N
6419921
CN(C)CCC=C1C2=CC=CC=C2COC3=CC=CC=C31.Cl
C19H22ClNO
MFCD00079135
doxepin hydrochloride, novoxapin, aponal, toruan, curatin, sinequan, zonalon, adapin, doxepine hydrochloride, novoxapin hydrochloride
(3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H361
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.

P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c

H301-H361

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 214-966-8

RTECSNumber : HQ4375000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only