missing translation for 'onlineSavingsMsg'
Learn More

Doxazosin Mesylate 98.0+%, TCI America™

Catalog No. p-7128113
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4126100MG 100 mg
D41261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4126100MG Supplier TCI America Supplier No. D4126100MG

Chemical Identifiers

CAS 77883-43-3
Molecular Formula C24H29N5O8S
Molecular Weight (g/mol) 547.583
MDL Number MFCD00216023
InChI Key VJECBOKJABCYMF-UHFFFAOYSA-N
Synonym doxazosin mesylate, cardura, carduran, cardular, cardura xl, cardenalin, dedralen, diblocin, doxazomerck, progandol
PubChem CID 62978
ChEBI CHEBI:4709
IUPAC Name [4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-(2,3-dihydro-1,4-benzodioxin-3-yl)methanone;methanesulfonic acid
SMILES COC1=C(C=C2C(=C1)C(=NC(=N2)N3CCN(CC3)C(=O)C4COC5=CC=CC=C5O4)N)OC.CS(=O)(=O)O

Specifications

Melting Point 269°C
Color White-Yellow
Quantity 100 mg
Formula Weight 547.58
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Doxazosin Mesylate
RTECSNumber TK8044000
TSCA No