missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dowtherm(R) SR-1, Spectrum™ Chemical
Specifications
Specifications
| CAS | 107-21-1, 7732-18-5, 7758-11-4 |
| Boiling Point | 163°C |
| Packaging | Steel Can |
| Quantity | 4 L |
| Grade | Reagent |
| Physical Form | Liquid |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.