missing translation for 'onlineSavingsMsg'
Learn More

Dowtherm(R) SR-1, Spectrum™ Chemical

Catalog No. 18610316
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610316 4 L Steel Can
18610314 20 L Steel Can
18610315 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610316 Supplier Spectrum Chemical Mfg Cor Supplier No. D12644LTGL
Only null left

Specifications

CAS 107-21-1, 7732-18-5, 7758-11-4
Boiling Point 163°C
Packaging Steel Can
Quantity 4 L
Grade Reagent
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.