missing translation for 'onlineSavingsMsg'
Learn More

Dowtherm(R) SR-1, Spectrum™ Chemical

Catalog No. 18610315
Change view
Click to view available options
Quantity:
20 L
200 L
Packaging:
Steel Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610315 200 L Steel Can
18610314 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18610315 Supplier Spectrum Chemical Mfg Cor Supplier No. D1264200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

Specifications

CAS 107-21-1, 7732-18-5, 7758-11-4
Boiling Point 163°C
Packaging Steel Can
Quantity 200 L
Grade Reagent
Physical Form Liquid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.