Learn More
AmberChrom™, 50WX8 200-400 (H)
CAS: 11119-67-8
Supplier: Thermo Scientific Chemicals L139220B
Description
AmberChrome™ 50WX8 Ion Exchange Resin) is used in separation medium for TLC and LPLC. It is also used as stripper resin and can be regenerated by 0.25N Sulfuricacid when exhausted, is used as catalyst and in deionizing supernatant fraction samples for paper chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmberChrome™ 50WX8 Ion Exchange Resin is used in separation medium for TLC and LPLC. It is also used as stripper resin and can be regenerated by 0.25N Sulfuricacid when exhausted, is used as catalyst and in deionizing supernatant fraction samples for paper chromatography.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
| Dowex™ 50WX8 200-400 (H) | |
| MFCD00132726 | |
| Insoluble in water. |
| 11119-67-8 | |
| 1000 g | |
| AmberChrom⢠50WX8 Ion Exchange Resin |
Chemical Identifiers
| 11119-67-8 | |
| MFCD00132726 | |
| AmberChrom⢠50WX8 Ion Exchange Resin |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P280-P305+P351+P338-P310
H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only