missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™, 50WX8 200-400 (H)

Catalog No. p-7046312
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1392218 50 g
AAL1392230 250 g
AAL139220B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1392218 Supplier Thermo Scientific Chemicals Supplier No. L1392218
Only null left

CAS: 11119-67-8

AmberChrome™ 50WX8 Ion Exchange Resin) is used in separation medium for TLC and LPLC. It is also used as stripper resin and can be regenerated by 0.25N Sulfuricacid when exhausted, is used as catalyst and in deionizing supernatant fraction samples for paper chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
AmberChrome™ 50WX8 Ion Exchange Resin is used in separation medium for TLC and LPLC. It is also used as stripper resin and can be regenerated by 0.25N Sulfuricacid when exhausted, is used as catalyst and in deionizing supernatant fraction samples for paper chromatography.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
IUPAC Name AmberChrom™ 50WX8 Ion Exchange Resin

Specifications

Quantity 50 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 50WX8 200-400 (H)
Hazard Category H318
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P280-P305+P351+P338-P310
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only