Learn More
Globe Scientific Double Tab Snap Caps for Blood Drawing Tubes, Glass and Plastic Tubes
Colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes and plastic culture tubes.
Supplier: Globe Scientific 113140G
Description
- Precision molded from LDPE
- Comes with two thumb tabs for easy removal of cap
- Provides tight seal during centrifugation, agitation and refrigeration procedures
- Protects samples from cross-contamination and highly infectious aerosols
- Color-coded for easy sample identification
- Color: green
Specifications
| 2 thumb tabs | |
| LDPE | |
| 13 mm Tubes |
| Snap Cap | |
| Green |