Learn More
Fisherbrand™ Double-Dipped PVC Gloves, Gauntlet Style
Provides enhanced cut resistance and excellent wet or slippery grip
$66.25 - $66.25
Specifications
Certifications/Compliance FDA 510(k)
Certifications/Compliance (Europe) CE marked
Applications Industrial
Latex Free Yes
Material PVC
View More Specs
Products 1
Catalog Number Mfr. No. Color Length (English) Grip Design Liner Material Price Quantity  
Catalog Number Mfr. No. Color Length (English) Grip Design Liner Material Price Quantity  
19-046-385
View Documents
Fisherbrand™
588030DD
Black 12 in. Rough Jersey
Dozen of 12 Pair for $66.25
Case of 6 Dozen for $397.50
Only null left
Description
Description
Gauntlet length, covers forearm
  • Available with sandy or rough acid grip finish
  • Enhanced cut resistance
  • Excellent wet or slippery grip
  • Interlock lining provides great dexterity
  • Jersey lining provides comfort and moderate insulation
Specifications
Specifications
FDA 510(k)
Industrial
PVC
Universal
Mining, construction, refining, petrochemicals and handling oily materials
Lined
CE marked
Yes
Chemical-resistant Gloves
EN 388:4121
Gauntlet
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'