missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Double Cylinder Hand Truck

Justrite™ Double Cylinder Hand Truck holds gas cylinders in place independently for improved safety. Features steel wheels with dual ball bearings for easier rolling.

Supplier:  Justrite™ 35028

 View more versions of this product

Catalog No. 19085759


Only null left

Description

Description

  • Accepts two gas cylinders with combined diameters up to 23-in (584-mm), including fuel and oxidizer cylinders common in welding applications
  • Constructed of 1-in (25-mm) and 1.25-in (32-mm) diameter round steel tube with continuous welding
  • Double-cylinder trucks provide optimum safety, strength, and longevity
  • Floor plate made of thick 11-gauge (3-mm) steel with safety fins prevents warping or breaking
  • Unique zero-lift height loading floor plate lays flat, thereby eliminating the need to lift cylinder during loading and unloading
  • Corrosion-resistant powder coat finish throughout for long life and smooth handling
  • Heavy-duty 1.5- x 54-in (38-mm x 1372-mm) polypropylene support straps, lab tested to 1138-lb (516-kg), feature steel cinch buckles for secure attachment of cylinder and eliminate any cylinder rolling potential seen in chain-secured units
  • Exclusive bumper guard edging grips cylinder and minimizes damage to cylinder and hand truck
  • Optional locking posts and collar plate system model 35084 ensures total cylinder immobilization, or accessorize hand trucks with a tool tray or lockable tool box for securing torches and other equipment

Compliance

OSHA, CGA P1, NFPA55

Warranty

1 YEAR

Specifications

Chemical Identifiers

Steel
Gray
Gas cylinder
1194 mm

Specifications

700 lb.
Steel
Gas cylinder
10.5 in. Pneumatic Wheels
23 lb.
318 kg
Gray
1194 mm
838 mm
838 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions