missing translation for 'onlineSavingsMsg'
Learn More

Dotriacontane 96.0+%, TCI America™

Catalog No. p-7126546
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09985G 5 g
D099825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D09985G Supplier TCI America Supplier No. D09985G
Only null left

Chemical Identifiers

CAS 544-85-4
Molecular Formula C32H66
Molecular Weight (g/mol) 450.88
MDL Number MFCD00009411
InChI Key QHMGJGNTMQDRQA-UHFFFAOYSA-N
Synonym n-dotriacontane, bicetyl, unii-7ksv90rn23, dicetyl, lacceran, dotriacontane, n, dotriacontane, acmc-1akq8, 4-01-00-00595 beilstein handbook reference, wln: 32h
PubChem CID 11008
ChEBI CHEBI:36020
IUPAC Name dotriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 70°C
Color White
Boiling Point 220°C
Quantity 5 g
Formula Weight 450.88
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dotriacontane
RTECSNumber JT2360000
TSCA Yes