missing translation for 'onlineSavingsMsg'
Learn More

Dotriacontane, 98%

Catalog No. p-7023678
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1748006 5 g
AAA1748014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1748006 Supplier Thermo Scientific Chemicals Supplier No. A1748006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 544-85-4 | C32H66 | 450.88 g/mol

Dotriacontane is used as a solvent medium for crystallization of isotactic polypropylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dotriacontane is used as a solvent medium for crystallization of isotactic polypropylene.

Solubility
Soluble in hot toluene, diethyl ether, benzene and ethanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 544-85-4
Molecular Formula C32H66
Molecular Weight (g/mol) 450.88
MDL Number MFCD00009411
InChI Key QHMGJGNTMQDRQA-UHFFFAOYSA-N
Synonym n-dotriacontane, bicetyl, unii-7ksv90rn23, dicetyl, lacceran, dotriacontane, n, dotriacontane, acmc-1akq8, 4-01-00-00595 beilstein handbook reference, wln: 32h
PubChem CID 11008
ChEBI CHEBI:36020
IUPAC Name dotriacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 68°C to 71°C
Density 0.812
Boiling Point 300°C to 310°C
Quantity 5 g
Beilstein 1783260
Solubility Information Soluble in hot toluene,diethyl ether,benzene and ethanol. Insoluble in water.
Formula Weight 450.88
Percent Purity 98%
Chemical Name or Material Dotriacontane
EINECSNumber 208-881-5
RTECSNumber JT2360000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only