missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ DOT Vehicle Placards: (Misc. Hazardous Materials w/Blank Box) 9
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
$4.38 - $16.55
Specifications
| Color | Black, White |
|---|---|
| Format | Label |
| Sterility | Non-sterile |
| Display Text | Misc. Hazardous Materials with blank box, 9 |
| For Use With (Application) | Material Handling, Shipping and Receiving Signs and Labels |
Description
- Available in two different materials Polycoated Tagstock and Premium Fiberglass
Specifications
| Black, White | |
| Non-sterile | |
| Material Handling, Shipping and Receiving Signs and Labels | |
| Diamond | |
| 10.75 in. |
| Label | |
| Misc. Hazardous Materials with blank box, 9 | |
| 1 Ea. | |
| 10.75 x 10.75 in. |