missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ DOT Vehicle Placards: (Misc. Hazardous Materials w/Blank Box) 9
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
Supplier: Brady™ BRD60404
Description
- Available in two different materials Polycoated Tagstock and Premium Fiberglass
Chemical Identifiers
| Black, White | |
| Non-sterile | |
| Material Handling, Shipping and Receiving Signs and Labels | |
| Diamond | |
| 10.75 in. |
| Label | |
| Misc. Hazardous Materials with blank box, 9 | |
| 1 Ea. | |
| 10.75 x 10.75 in. |
Specifications
| Non-adhesive | |
| Semi-gloss | |
| Polycoated Tagstock (B-101) | |
| Vehicle Placard | |
| Misc. Hazardous Materials with blank box, 9 | |
| 1 Ea. | |
| 10.75 x 10.75 in. | |
| 10.75 x 10.75 in. |
| Black, White | |
| Label | |
| Single-side Printable | |
| Non-sterile | |
| Material Handling, Shipping and Receiving Signs and Labels | |
| Diamond | |
| 0.0024 in. | |
| 10.75 in. |