missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ DOT Vehicle Placards: FLAMMABLE SOLID 4
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
$16.30 - $16.30
Specifications
| Color | Black, Red, White |
|---|---|
| Product Type | Vehicle Placard |
| Display Text | FLAMMABLE SOLID 4 |
| For Use With (Application) | Material Handling, Shipping and Receiving Signs and Labels |
| Quantity | 1 Ea. |
Description
- Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Specifications
| Black, Red, White | |
| FLAMMABLE SOLID 4 | |
| 1 Ea. | |
| 10.75 x 10.75 in. |
| Vehicle Placard | |
| Material Handling, Shipping and Receiving Signs and Labels | |
| Diamond |