missing translation for 'onlineSavingsMsg'
Learn More

Brady™ DOT Vehicle Placards: FLAMMABLE 3

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

$4.38 - $17.96

Specifications

Color White, Red
Format Label
Sterility Non-sterile
Display Text FLAMMABLE 3
For Use With (Application) Material Handling, Shipping and Receiving Signs and Labels
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19111323
View Documents Promotion Details
Brady™
63409
Pressure-sensitive Vinyl (B-946)
Each for $17.96
Only null left
 
19111347
View Documents Promotion Details
Brady™
63443
Polycoated Tagstock (B-101)
Each for $4.38
Only null left
 
Description

Description

  • Available in two different materials Pressure Sensitive Vinyl, Polycoated Tagstock
Specifications

Specifications

White, Red
Non-sterile
Material Handling, Shipping and Receiving Signs and Labels
Diamond
10.75 in.
Label
FLAMMABLE 3
1 Ea.
10.75 x 10.75 in.
Videos
SDS
Documents

Documents