missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ DOT Vehicle Placards: EXPLOSIVE 1.1D
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
$4.32 - $4.32
Specifications
| Color | Black, Orange |
|---|---|
| Product Type | Vehicle Placard |
| Display Text | EXPLOSIVE 1.1D |
| For Use With (Application) | Material Handling, Shipping and Receiving Signs and Labels |
| Quantity | 1 Ea. |
| Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
|
19-111-299
|
Brady™
63373 |
Polycoated Tagstock |
Each for $4.32
|
|
|||||
Description
- Available in two different materials Polycoated Tagstock, Pressure Sensitive Vinyl
Specifications
| Black, Orange | |
| EXPLOSIVE 1.1D | |
| 1 Ea. | |
| 10.75 x 10.75 in. |
| Vehicle Placard | |
| Material Handling, Shipping and Receiving Signs and Labels | |
| Diamond |