missing translation for 'onlineSavingsMsg'
Learn More

Brady™ DOT Vehicle Placards: EXPLOSIVE 1.1D

Catalog No. 19111299
Change view
Click to view available options
Material:
Polycoated Tagstock
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19111299 Polycoated Tagstock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19111299 Supplier Brady™ Supplier No. 63373

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

  • Available in two different materials Polycoated Tagstock, Pressure Sensitive Vinyl

Specifications

Adhesive Type Non-adhesive
Color Black, Orange
Finish Semi-gloss
Format Label
Material Polycoated Tagstock
Product Type Vehicle Placard
Sterility Non-sterile
Display Text EXPLOSIVE 1.1D
For Use With (Application) Material Handling, Shipping and Receiving Signs and Labels
Quantity 1 Ea.
Shape Diamond
Size (English) 10.75 x 10.75 in.
Thickness (English) 0.0024 in.
Dimensions (L x W) 10.75 x 10.75 in.
Length (English) 10.75 in.
Width (English) 10.75 in.
Show More Show Less