Learn More
METTLER TOLEDO™ Dosing Head Accessory
Catalog No. 01676965
Change view
Click to view available options
Product Line:
Dosing Head Liquid QL001
Powder Dosing Head QH002-CNMW
Powder Dosing Head QH008-BNMP
Powder Dosing Head QH010-CNMW
Powder Dosing Head QH012-LNMX
QH012-LNJW Powder Dosing Head
QL003 Liquid Dosing Head
Quantity:
1 Ea.
10 Pieces
150 Pieces
40 Pieces
500 Pieces
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line Quantity
01676965 QL003 Liquid Dosing Head 1 Ea.
01677010 QH012-LNJW Powder Dosing Head 10 Pieces
01677017 - 1 Ea.
01677018 Powder Dosing Head QH010-CNMW 40 Pieces
01677019 Powder Dosing Head QH010-CNMW 10 Pieces
01677020 Powder Dosing Head QH012-LNMX 10 Pieces
01677021 Dosing Head Liquid QL001 1 Ea.
01677022 Powder Dosing Head QH010-CNMW 150 Pieces
01677033 Powder Dosing Head QH008-BNMP 10 Pieces
01677014 Powder Dosing Head QH002-CNMW 500 Pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 01676965 Supplier METTLER TOLEDO™ Supplier No. 30542814
Only null left
Dosing accessories efficiently dispense accurate samples for high throughput dosing activities.
  • Automated dispensing in one unit
  • Efficient preparation of accurate samples
  • Built-in RFID chip
Specifications
For Use With (Equipment) Excellence XPR Automatic Balance
Quantity 1 Ea.
Product Line QL003 Liquid Dosing Head
Type Liquid Dosing Head