missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Sheep, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP184PMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP184PMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP184PMI Supplier MilliporeSigma Supplier No. AP184P

Donkey Polyclonal Antibody

Detect Donkey Sheep IgG using this Donkey anti-Sheep IgG Antibody, HRP conjugate, Species Adsorbed validated for use in ELISA and western blotting.

Specifications

Antigen Donkey anti-Sheep IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Sheep

For Research Use Only