missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Sheep, Cy5, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP184SMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP184SMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP184SMI Supplier MilliporeSigma Supplier No. AP184S
Only null left

Donkey Polyclonal Antibody

Donkey anti-Sheep IgG Antibody, Cy5 conjugate, Species Adsorbed detects level of Donkey Sheep IgG and has been published and validated for use in immunofluoresence.

Specifications

Antigen Donkey anti-Sheep IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Conjugate Cyanine5
Host Species Donkey
Quantity 500 μg
Primary or Secondary Secondary
Target Species Sheep

For Research Use Only