missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Sheep, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP184AMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP184AMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP184AMI Supplier MilliporeSigma Supplier No. AP184A

Donkey Polyclonal Antibody

Donkey anti-Sheep IgG Antibody, Alkaline Phosphatase conjugate, Species Adsorbed detects level of Donkey Sheep IgG and has been published and validated for use in ELISA and western blotting.

Specifications

Antigen Donkey anti-Sheep IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.6mg/mL
Conjugate Alkaline Phosphatase (AP)
Formulation Reconstitute with sterile distilled water to match the volume indicated on the vial label. Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature.
Host Species Donkey
Purification Method Affinity Purified
Quantity 500 μL
Primary or Secondary Secondary
Target Species Sheep
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only