missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Rat, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP189PMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP189PMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP189PMI Supplier MilliporeSigma Supplier No. AP189P

Donkey Polyclonal Antibody

Donkey anti-Rat IgG Antibody, HRP conjugate, Species Adsorbed detects level of Donkey Rat IgG and has been published and validated for use in ELISA and western blotting.

Specifications

Antigen Donkey anti-Rat IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Rat

For Research Use Only