missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Rat, FITC, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP189FMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP189FMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP189FMI Supplier MilliporeSigma Supplier No. AP189F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Donkey Polyclonal Antibody

This Donkey anti-Rat IgG Antibody, FITC conjugate, Species Adsorbed is validated for use in IF for the detection of Donkey Rat IgG.

Specifications

Antigen Donkey anti-Rat IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Concentration 1.5mg/mL
Conjugate FITC
Formulation Freeze-dried powder.
Host Species Donkey
Immunogen Highly purified Rat IgG isolated from pooled normal Rat Serum.
Quantity 500 μg
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C. Do not freeze.
Form Freeze-dried
Show More Show Less

For Research Use Only