missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Rat, Cy3, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP189CMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP189CMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP189CMI Supplier MilliporeSigma Supplier No. AP189C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Donkey Polyclonal Antibody

Donkey anti-Rat IgG Antibody, Cy3 conjugate, Species Adsorbed is an antibody against Donkey Rat IgG for use in immunofluoresence.

Specifications

Antigen Donkey anti-Rat IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Conjugate Cyanine3
Host Species Donkey
Quantity 500 μg
Primary or Secondary Secondary
Target Species Rat

For Research Use Only