missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Donkey anti-Rabbit, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™
Description
Donkey anti-Rabbit IgG Antibody, Rhodamine conjugate, Species Adsorbed is an antibody against Donkey Rabbit IgG for use in immunofluoresence.
Spécifications
Spécifications
| Antigen | Donkey anti-Rabbit IgG Species adsorbed |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Rhodamine |
| Formulation | Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, and 0.05% sodium azide. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water. RHODAMINE/PROTEIN: Approximately 0.52 (A550/A280) |
| Host Species | Donkey |
| Quantity | 500 μg |
| Primary or Secondary | Secondary |
| Target Species | Rabbit |
| Content And Storage | 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. |
| Afficher plus de résultats |
For Research Use Only