missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Rabbit, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP182PMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP182PMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP182PMI Supplier MilliporeSigma Supplier No. AP182P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Donkey Polyclonal Antibody

Donkey anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed is an antibody against Donkey Rabbit IgG for use in ELISA and western blotting.

Specifications

Antigen Donkey anti-Rabbit IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate HRP
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA. Contains no preservative. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water.
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only