Learn More
Donkey anti-Rabbit IgG (H+L) Secondary Antibody, WesternDot™ 625, Invitrogen™
Description
WesternDot fluorescently labeled secondary antibodies deliver superior signal-to-noise ratios and exhibit comparable sensitivity to enhanced chemiluminescence (ECL)-based methods in western blot applications. No stripping and re-probing or multiple blots are required for multiple protein detection as commonly needed with ECL or colorimetric detection. Differently labeled WesternDot antibodies can be applied to a single blot for simultaneous detection of multiple proteins using standard gel or blot imaging platforms (a list of instruments that are compatible with WesternDot detection can be found in the user manual below). Our WesternDot antibodies utilize Qdot nanocrystals enhanced with VIVID technology, making them brighter than the original Qdot reagents. Additionally, they have reduced intensity differences between the different colors, simplifying multiplex applications.
Specifications
Specifications
| Antigen | Rabbit IgG (H+L) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | WesternDot 625 |
| Formulation | 50mM borate with 1M betaine and 0.05% sodium azide; pH 8.3 |
| Host Species | Donkey |
| Immunogen | Rabbit Immunoglobins |
| Purification Method | purified |
| Quantity | 25 blots |
| Regulatory Status | RUO |
| Show More |