missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP
Description
Product SA1-200 is conjugated to HRP and optimized for ECL applications. This antibody reacts with the heavy and light chains of the immunoglobulin molecule. Pre-absorbed with the following species proteins: Bovine, chicken, goat, guinea pig, syrian hamster, horse, human, mouse, rat, and sheep serum proteins.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Rabbit IgG (H+L) Cross-Adsorbed |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 40 μg/mL |
| Conjugate | HRP |
| Formulation | PBS with proprietary stabilizer, 50% glycerol and 0.07% Kathon |
| Host Species | Donkey |
| Purification Method | Antigen affinity chromatography |
| Quantity | 250 μL |
| Regulatory Status | RUO |
| Show More |