Learn More
Invitrogen™ Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31821 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31821 reacts with the heavy chains of rabbit IgG and with the light chains common to most rabbit immunoglobulins, but does not react against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rat and sheep serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 0.5 mL of distilled water and centrifuge if not clear. Country of Origin: USA.
Specifications
Specifications
| Antigen | Rabbit IgG (H+L) Cross-Adsorbed |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 |
| Host Species | Donkey |
| Purification Method | Antigen affinity chromatography |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |