missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Rabbit, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP182BMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP182BMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP182BMI Supplier MilliporeSigma Supplier No. AP182B

Donkey Polyclonal Antibody

Donkey anti-Rabbit IgG Antibody, biotin-SP conjugate, Species Adsorbed detects level of Donkey Rabbit IgG and has been published and validated for use in ELISA and western blotting.

Specifications

Antigen Donkey anti-Rabbit IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.15M NaCl, pH 7.1 with 15mg/mL BSA, and 0.1% sodium azide. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water.
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only