missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Rabbit, Alkaline Phosphatase, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP182AMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP182AMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP182AMI Supplier MilliporeSigma Supplier No. AP182A

Donkey Polyclonal Antibody

This Donkey anti-Rabbit IgG Antibody, Alkaline Phosphatase conjugate, Species Adsorbed is validated for use in ELISA, western blotting for the detection of Donkey Rabbit IgG.

Specifications

Antigen Donkey anti-Rabbit IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alkaline Phosphatase (AP)
Host Species Donkey
Quantity 500 μL
Primary or Secondary Secondary
Target Species Rabbit

For Research Use Only