missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Mouse, Rhodamine (TRITC), Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP192RMI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP192RMI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP192RMI Supplier MilliporeSigma Supplier No. AP192R

Donkey Polyclonal Antibody

Donkey anti-Mouse IgG Antibody, Rhodamine conjugate, Species Adsorbed is an antibody against Donkey Mouse IgG for use in immunofluoresence.

Specifications

Antigen Donkey anti-Mouse IgG Species adsorbed
Applications Immunofluorescence
Classification Polyclonal
Conjugate Rhodamine
Formulation Lyophilized. Buffer = 0.02M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA, and 0.1% sodium azide. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water. RHODAMINE/PROTEIN: Approximately 0.5 (A550/A280).
Host Species Donkey
Quantity 500 μg
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Lyophilized
Show More Show Less

For Research Use Only