missing translation for 'onlineSavingsMsg'
Learn More

IgG Donkey anti-Mouse, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP192PMI
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP192PMI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP192PMI Supplier MilliporeSigma Supplier No. AP192P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Donkey Polyclonal Antibody

Donkey anti-Mouse IgG Antibody, HRP conjugate, Species Adsorbed is an antibody against Donkey Mouse IgG for use in ELISA and western blotting.

Specifications

Antigen Donkey anti-Mouse IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate HRP
Formulation Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.15M NaCl, pH 7.1 with 15mg/mL BSA. RECONSTITUTION: Reconstitute with 500μL of sterile distilled water.
Host Species Donkey
Purification Method Immunoaffinity Purified and Immunodepletion
Quantity 500 μL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only