missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Donkey anti-Mouse IgG (H+L) Secondary Antibody, TRITC
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
| Antigen | Mouse IgG (H+L) |
| Applications | Dot Blot, ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.2 mg/mL |
| Conjugate | TRITC |
| Formulation | PBS with 1% BSA and 0.01% sodium azide; pH 7.2 |
| Host Species | Donkey |
| Immunogen | Mouse IgG whole molecule. |
| Purification Method | IgG fraction |
| Quantity | 1 mg |
| Show More |