Learn More
Invitrogen™ Donkey anti-Mouse IgG (H+L) Secondary Antibody, AP
Donkey Polyclonal Secondary Antibody
Supplier:  Invitrogen™ PA128749
Catalog No. PA128749
Only null left
Explore available promotions
Description
Description
PA1-28749 detects Mouse IgG from mouse samples.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Mouse IgG (H+L)
Polyclonal
Alkaline Phosphatase (AP)
Donkey
Antigen affinity chromatography
RUO
Mouse
Secondary Antibody
IgG
Dot Blot, ELISA, Immunohistochemistry, Western Blot
1 mg/mL
TBS with 1% BSA, 0.001M MgCl2, 0.001M ZnCl2, 50% glycerol and 0.1% sodium azide; pH 8.0
Mouse IgG whole molecule.
100 μg
Secondary
4°C, do not freeze
Liquid
Product Suggestions
Product Suggestions