missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594

Catalog No. A21203
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21203 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A21203 Supplier Invitrogen™ Supplier No. A21203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Donkey Polyclonal Secondary Antibody

These donkey anti-mouse IgG (H+L) whole secondary antibodies have been affinity-purified and show minimum cross-reactivity to bovine, chicken, goat, guinea pig, hamster, horse, human, rabbit, rat, and sheep serum proteins. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g., flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 594 dye is a bright, red-fluorescent dye with excitation ideally suited to the 594 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 594 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of low-abundance biological structures with great sensitivity. Alexa Fluor 594 dye molecules can be attached to proteins at hi...

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Mouse IgG (H+L) Highly Cross-Adsorbed
Applications Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Alexa Fluor 594
Formulation PBS with 5mM sodium azide; pH 7.5
Host Species Donkey
Immunogen Gamma Immunoglobins Heavy and Light chains.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C, store in dark
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less